Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Glo Germ
Supplier: GLO GERM SE
GEL POWDER BRHP OIL
470100-620EA 25.5 USD InStock
470100-620 470004-262 470004-260 470100-682
Glo Germ
Educational Materials Biology Educational Materials Physiology Learning Activities
Used Worldwide to Teach Proper Handwashing Technique.
  • Choose Powder, Liquid, or Gel
  • Test Handwashing or Cleaning Effectivity
  • Invisible Until Exposed to UV Light
Both the oil and the powder contain minute pieces of plastic that glow under UV light. To become germ detectives, students apply a small amount of oil before handwashing and then try to spot remaining “germs” under a UV light. To investigate the thoroughness of surface cleaning, students wipe away a light dusting of Glo Germ powder and hunt for germs with the UV light.
Glo Germ gel is recommended where stains are of concern. The adjustable flow Glo Brush lets users apply a thin layer of powder to any surface quickly and without waste and features a refillable powder reservoir with a cover to protect bristles.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty