Would you like to log in again?
CENCO® Mechanical Oscillator
Supplier: STEINER ENTERPRISES SE
CP36803-01
470004-272EA 297.95 USD InStock
470004-272
CENCO® Mechanical Oscillator
Educational Materials Physics Educational Materials Waves and Oscillations Learning Activities
The Mechanical Oscillator is perfect for performing existing harmonic motion demonstrations or creating new ones.
  • Oscillator Provides Mechanical Vibrations for Driving Strings, Springs, and More
  • Range: 0 - 50 kHz
  • Fuse-Protected Input
  • Rubber Bumpers for Vertical and Horizontal Positioning
The oscillator provides mechanical vibrations for driving strings, springs, ripple tanks, Chladni Plates, or other experiments. The input is fuse protected to prevent inadvertent damage. Rubber bumpers under the base and on one side allow for both vertical and horizontal positioning. The base can be mounted on a standard 1.3 cm rod in the vertical and horizontal direction. Amplitude up to 7 mm (decreases as frequency increases).
Connect accessories by plugging them into 4 mm horizontal and vertical holes in the drive shaft. Instructions are included. A Oscillator Amplifier and a Function Generator are required but not included.
Power requirements: Provide 1 Amp to a 6 ohm impedance
Ordering information: Instructions included
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty