Would you like to log in again?
WaterWorks™ School Test Kit
Supplier: INDUSTRIAL TEST SYSTEMS INC SE
487942 487995
470004-284EA 161.7 USD InStock
470004-284 470151-358
WaterWorks™ School Test Kit
Educational Materials Earth Science Educational Materials Natural Resources Learning Activities
The WaterWorks School Test Kit is a great introduction to water-quality testing.
  • Safe, easy-to-use strips
  • Nine different tests
  • Enough material to test each factor 60 times
  • Manual included
  • Great introduction to water-quality testing
These safe, easy-to-use test strips eliminate the need to mix and dispose of hazardous chemicals and handle glassware. You just dip a test strip in a water sample and then compare it to the included color chart to interpret the results. This kit contains tests for free chlorine, total chlorine, iron, copper, pH, nitrates, nitrites, total alkalinity, and total hardness: nine different tests in all.
It comes with enough material to test each factor 60 times, comprising a total of 540 tests to ensure that every student gets hands-on experience. It also includes a manual with background information on water quality and 11 activities on topics such as creating a water cycle, building a well, and testing for acid rain.
The 9-WAY Test kit tests for the same factors as the School test kit but contains 2 tests for each factor making it ideal for individual projects.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty