Would you like to log in again?
Basic Soil Sampler Set
Supplier: Avantor
WARD470004-344
470004-344KT 112.99 USD InStock
470004-344
Basic Soil Sampler Set
Educational Classroom Kits and Activities
Get Perfect Samples in Any Kind of Soil
  • Remove Your Core Intact
  • Easy to Use in the Field
  • Teacher Guide Included
Use the boring handle to select your sample, then just remove it and replace it with a special extractor unit. This special handle neatly and easily removes your soil plug intact. The sampler also features a depth measurement scale, allowing you to collect multiple samples from uniform depths up to 8" with a minimum of fuss. When you are finished in the field, just use the boring handle or a stiff brush to clean it. It comes with enough soil collection bags and labels for 30 students, a teacher’s guide, and a student study guide copymaster.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty