Would you like to log in again?
Reduce; Reuse; Recycle: A Plastics Identification Lab Activity
Supplier: Avantor
WARD470004-352
470004-352KT 116.99 USD InStock
470004-352
Reduce; Reuse; Recycle: A Plastics Identification Lab Activity
Educational Classroom Kits and Activities
Make Your Students “Plastic Literate”
  • Learn to differentiate between different plastics
  • Linked to real world scenarios
  • Includes instructions
Increase students’ awareness of plastics in everyday use. Activities include interpreting recycling symbols on plastic materials, and performing scientific tests (density, alcohol, etc.) to identify various types of plastics. Includes materials for 30 students and instructions.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty