Would you like to log in again?
Seasonal Demonstration Globe
Supplier: THE SCIENCE SOURCE SE
U020
470004-404EA 142 USD InStock
470004-404
Seasonal Demonstration Globe
Globes
Economical Example of Earth’s Rotation and Seasons
Observe the Earth, tilted on its axis, as it rests on a disk divided into months and days of the month. The globe is half in, half out of a black plexiglas box, representing day and night. To see what part of the planet is tilted toward the sun, indicating which areas are experiencing warmer summer months, simply rotate the globe and disk to line up the day you choose with the stationary plastic indicator on the base, or you can rotate the globe by itself to demonstrate day/night. The disk also features the constellations that are visible during each month. Globe size: 6" dia; plexiglas base size: 7"L x 7"W x 7 1/4"H.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty