Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Dogfish Dissection Tray
Supplier: NASCO
SB14284 SB14454DIM
470004-500EA 41.55 USD InStock
470004-500 470004-502
Dogfish Dissection Tray
Trays/Dishes/Pots Dissecting Trays
This lightweight, chemically resistant polyethylene tray holds even the largest dogfish specimen, ventral side up, because the molded central channel and grove hold the dorsal fin and spine in place.
It also features a nonskid bottom. Size: 31"L×151/2"W×15/8"H.
Ordering information: The clear plastic cover is available separately.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty