Would you like to log in again?
Bed of Nails Demonstration Model
Supplier: TONAWANDA MANUFACTURING
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
47418-00
470004-754EA 82.65 USD InStock
470004-754
Bed of Nails Demonstration Model
Educational Materials Physics Educational Materials Mechanics Learning Activities
Reveal the Physics of Pressure
Show your students the difference between force and pressure by applying force over a large area. The larger the area, the less likely the balloon is to break. It includes instructions and all materials but the balloons and masses.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty