Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
H-B® Easy Read® Environmentally Friendly, General Purpose Liquid-In-Glass Thermometers
Supplier: Bel-Art Products
B60302-0000_SE B60301-0200_SE B60301-0200_SE B60301-0300_SE
470004-838EA 30.4 USD InStock
470004-838 470123-626 470004-834 470202-822
H-B® Easy Read® Environmentally Friendly, General Purpose Liquid-In-Glass Thermometers
Thermometers Analog Thermometers
EnviroKleen™ certified thermometers feature black, non-toxic, biodegradable, Enviro-Safe® liquid against lead-free, yellow back glass for easy readability and fewer reading errors.
  • Non-toxic and biodegradable liquid
  • Lead-free
  • Yellow back glass
  • Easy to read
Ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries.
Ordering information: Manufactured and tested by our USA quality team, these thermometers are traceable to NIST and include a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy and Traceability. Should you require an individual Thermometer Calibration Report, factory calibrated thermometers or custom calibration services are available.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty