Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Digital Timing System with Photogates

Supplier: VWR International
LBTM
470004-892EA 562.6 USD InStock
470004-892
Digital Timing System with Photogates
Timers Digital Timers
Microprocessor-controlled digital timer has eight built-in measurement modes.

  • Suitable for use with free fall apparatus
  • Displays LED 14 mm
  • Voltage in 110 V

This is a precision digital timer used to measure the time interval between any two events with high accuracy. The timer standard accuracy has been achieved using the 10 MHz Quartz Crystal Oscillator. It has four operating modes manual, free fall mode, timing gates mode and any other triggering start and stop. It has a built in power source for timing gates and the coil for g by free fall.

Delivery information: Supplied with two timing gates and manual.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty