Would you like to log in again?
Precision Gyroscope
Supplier: TEDCO INC CP
30000
470004-966EA 17.85 USD InStock
470004-966
Precision Gyroscope
Educational Materials Physics Educational Materials Mechanics Learning Activities
This gyroscope will spin for several minutes.
  • Ready to use right out of the box
  • Unparalleled ease-of-use and performance
  • Use string or T-handle zip starter to initiate spinning
  • Simple and effective tool to teach kids about physics
This is the most precision designed toy gyroscope on the market. Tiny ball bearings and near perfect balance keep the gyroscope spinning for several minutes. The included T-handle zip starter strap drastically improves ease-of-use. Durable enough for classroom demonstration.
Delivery information: Includes precision gyroscope, pedestal, string and T-handle zip starter.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty