Would you like to log in again?
Equal Mass Density Cubes
Supplier: AJAX SCIENTIFIC LTD CP
ME055-0520
470004-982EA 24.1 USD InStock
470004-982
Equal Mass Density Cubes
Educational Materials Physics Educational Materials Mechanics Learning Activities
Set of 5 metal cubes of equal (20 g) mass. Cubes are copper, aluminum, zinc, iron and brass. Demonstrate inverse relationship between density and volume of equal mass metal cube.
Caution: CHOKING HAZARD - Small parts. Not for children under 3 yearrs.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty