Would you like to log in again?
Dr. Fuel Cell™ Basic Science Kit
Supplier: HELIOCENTRIS GMBH CP
K0350E K0356E 352E
470005-000EA 360.45 USD InStock
470005-000 470004-608 470004-996
Dr. Fuel Cell™ Basic Science Kit
Educational Materials Physics Educational Materials Alternative Energy Learning Activities
With this collection of science kits, you and your students can explore the concepts behind and the many applications of solar hydrogen, direct methanol, and other fuel cell technologies.
The Solar Hydrogen Kit contains over 20 different experiments that explore this emerging technology using the same techniques employed in advanced industrial applications. The Direct Methanol Fuel Cell lets students perform a wide range of experiments covering the working principles of the methanol fuel cell and the potential uses of this amazing energy source.
The Dissectible Fuel Cell expands the Hydrogen Science Kit, allowing for more experiments and an in-depth examination of the principles and process parameters of fuel cells. The Hydrogen Fuel Cell Model Car is for those who want to demonstrate a driving fuel cell car. THe kit includes a solar module to power the reversible fuel cell for charging-up the car with hydrogen and all necessary cables.
The kit with four manuals includes: PEM fuel cell, PEM-electrolyser (1W) with gas-storage cylinders, solar module, load measurement box, stop watch, connecting cable and silicone tubes, tubing stoppers, replacement bulb, and instruction booklet. Direct Methanol Fuel Cell includes methanol fuel cell with tank, load measurement box, three methanol bottles, funnel, spout, cables, storage container with lid and instructions.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty