Would you like to log in again?
Radon Test Kit
Supplier: AIR CHEK INC.
AIR CHECK
470005-098EA 42.8 USD InStock
470005-098
Radon Test Kit
Standards Environmental Analysis Standards
Monitors Hazardous Radon Levels
Produced by the natural breakdown of uranium, the odorless, colorless, tasteless gas, radon is known to cause lung cancer when found in high concentrations. This gas can be found in certain kinds of common rocks and soils and can be safely tested for with the contents of this kit. A charcoal sampling packet and a prepaid mailer are included so you can expose the packet to suspected radon gases, then seal and return the mailer to the test lab for analysis. The lab will return the test results (radon concentration in picocuries per liter), together with background and interpretative information. Note: kit is not available for sale in New Jersey.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty