Would you like to log in again?
Ward's® Aquarium Gravel
Supplier: INDUSTRIAL SUPPORT INC.
Ward's®
6024000 WARD470021-312
470021-312BX 46.5 USD InStock
470021-312 470005-160
Ward's® Aquarium Gravel
Animal Supplies Animal Environmental Enrichment
This decorative gravel is also essential for maintaining your aquarium, acting as a substrate for the bacteria cultures necessary for a healthy tank. Requires 1 lb. of gravel per gallon.
Attractive Bottom Cover Enhances Natural Filtration and Nitrogen Cycling
Gravel contains no lead or chrome and won’t change the pH or chemistry of the tank. Pebble sizes up to 1/4" dia.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty