Would you like to log in again?
Fine-Point Forceps With Guide Pins
Supplier: DUNRITE INSTRUMENTS SE
140541 140531
470005-440BX 209 USD InStock
470005-440 470007-560
Fine-Point Forceps With Guide Pins
Forceps
Slip proof fine point forceps.
  • Guide pins keep tip aligned
  • Prevents slipping and twisting when closed
  • Textured tips improve hold on slippery material
Used in our own labs, these forceps feature a built-in guide pin to keep the fine tips aligned, which is important for grasping individual vessels or nerves. Stainless steel. Length: 4¹/₂".
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty