Would you like to log in again?
Flint Glass Tubing
Supplier: GSC INTERNATIONAL SE
W00432 6MMSLT-24 10MMSLT-24-CS 5MMSLT-24
470235-726CS 261.95 USD InStock
470235-726 470005-532 470206-452 470005-534
Flint Glass Tubing
Tubing Transfer Tubing
Perfect Laboratory Glass Tubing.
  • Sold by the lb.
  • All lengths 60 cm
Made from fine quality flint glass for easy bending, blowing, and sealing of soft glass apparatus. Wall uniformity controlled within ±0.05 mm. Tolerance of the outside diameter is ±0.20 mm.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty