Would you like to log in again?
Meter Ecology Activity
Supplier: Avantor
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
WARD470005-542
470005-542EA 88.99 USD InStock
470005-542
Meter Ecology Activity
Educational Classroom Kits and Activities
Ideal to perform vegetation and organism studies to assess the lifeforms growing in an area.
In this field ecology activity, students will use a meter quadrant square and surveyor flags to stake out one square meter of ground in up to seven locations, and then examine the insects and other organisms found there. The groups share information, compare differences, and identify like organisms within each quadrant. Students can also perform leaf litter studies. Developed by Deborah Mann, this activity includes materials for seven setups and age-appropriate instructions for grades 4–7, 8–10 and 11–12.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty