Would you like to log in again?
Bibulous Paper
Supplier: RAYLABCON INC SE
Avg. Ratings: 5.0 (1 Review)
REVIEWS (1)
Total Ratings: 1
Avg. Ratings: out of 5
Sort all by:
20105000
470005-600PK 6.15 USD InStock
470005-600
Bibulous Paper
Wipes Disposable Wipes and Towels Lens Papers
Common blotting and cleaning paper.
  • Quality slide making supplies
  • Helpful for maintaining cleanliness of slides
Use this bibulous paper to clean glass surfaces of dust and lint without leaving streaks. It also blots excess moisture from slides containing living materials. Staple-bound booklet of 50 sheets.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (1)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty