Would you like to log in again?
Wooden Meter and Half-Meter Sticks
Supplier: AMERICAN SCIENTIFIC LLC SE
2192-98 104905
470017-002EA 6.5 USD InStock
470017-002 470005-628
Wooden Meter and Half-Meter Sticks
Rulers
Good quality hardwood meter and half-meter sticks with a natural finish.
  • Permanently engraved scales
  • One side is graduated in cm with 1mm divisions
  • Other side is graduated in inches with 1/8-inch divisions
Meter sticks are 6mm thick x 25.4mm wide.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty