Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Solar Spectrum Chart
Supplier: WABASH INSTRUMENTS CORP SE
SP-188 332326
470173-990EA 33.39 USD InStock
470173-990 470005-830
Solar Spectrum Chart
Charts
Depicts the Most Prominent Fraunhofer Lines and Their Wavelengths
  • Easy-to-read
  • Heavyweight
  • Available in two sizes
This poster shows the most prominent Fraunhofer lines along with their wavelength and letter assignment. The chart identifies the chemical elements in the sun’s atmosphere represented by the lines. Features metal edges at top and bottom and sturdy metal loop hangers.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty