Would you like to log in again?
High RPM Steam Engine
Supplier: JENSEN MFG CO INC SE
70
470005-846EA 356.85 USD InStock
470005-846
High RPM Steam Engine
Educational Materials Physics Educational Materials Heat/Thermodynamics Learning Activities
The High rpm Steam Engine features an electrically-heated horizontal boiler.
  • Water-Gauge Safety Valve
  • Whistle
  • Throttle
  • Two-Speed Pulley
  • Electronically-Heated Horizontal Boiler
The apparatus includes a water-gauge safety valve, whistle, throttle, two-speed pulley, and 400W heater.
The engine may be used to drive our Electric Generator.
Ordering information: The engine includes a three-wire cord and plug for operation on 110VAC.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty