Would you like to log in again?
Flint Glass Watch Glasses
Supplier: United Scientific Supplies
WGL090 WGL065 WGL050
470005-916PK 31.5 USD InStock
470005-916 470104-600 470020-744
Flint Glass Watch Glasses
Watch Glasses
Useful clear watch glasses for evaporation.
  • Flint glass
  • Flame polished edges
Made from clear flint glass (1.8 to 2.0mm thick), fine ground and polished edges.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty