Would you like to log in again?
Denoyer-Geppert® Comprehensive Ear Model
Supplier: DENOYER-GEPPERT SCI CO SE
0123-00
470005-984EA 924.95 USD InStock
470005-984
Denoyer-Geppert® Comprehensive Ear Model
Models Human Biology Models
Removable Parts Show Structure Within Temporal Bone
  • Available With, Or Without Bypass
  • Made Of Highly Break Resistant Plastic
Study the anatomy of the ear from a different perspective. Unlike other models that show a temporal bone sectioned along the auditory canal, this model features removable sections of the temporal bone to clearly show the pathways of sound reception. In addition, the transparent, liquid-filled semicircular canals demonstrate the process of maintaining equilibrium. The skin is removed from the pinna to show the cartilage, nerves, vessels, and muscles and the pinna is removable for closer study. The petrous is sectioned horizontally and can be removed to reveal the middle and inner ear chambers; the mastoid can be detached to show the entire middle ear. The tympanum with malleus, incus, and stapes, as well as the two-part cochlea showing how the vestibular and cochlear nerves join to form the auditory nerve are all removable as well. The eight-part model is mounted on a base.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty