Would you like to log in again?
Motorized, Illuminated Planetarium
Supplier: SCIENCE INTERACTIVE GROUP
653-3020
470006-098EA 1231 USD InStock
470006-098
Motorized, Illuminated Planetarium
Models Astronomy Models
Completely Automated Instrument with Synchronized Orbits
  • Model has a built in compass
  • Sun provides illumination
Demonstrate the motions of the Earth, moon, sun, and Venus. The model features synchronized orbital movements for the sun, Earth, and Venus; illuminated 6"-diameter sun; and a built-in compass. 115 VAC with a 5' cord. Length: 16 1/2".
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty