Would you like to log in again?
Kemtec® Photosynthesis Reaction Activities
Supplier: AQUAPHOENIX SCIENTIFIC INC. SE
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
2-516 2-517
470007-840KT 168.4 USD InStock
470007-840 470006-236
Kemtec® Photosynthesis Reaction Activities
Educational Materials Biology Educational Materials Plant Learning Activities
Two activities to explore photosynthesis.
  • Examine photosynthesis in two parts
  • Examine one part or both
Photosynthesis I examines the hill reaction in isolated chloroplasts; splitting water to yield oxygen and release hydrogen. The proteolysis of water in the course of the reaction is measurable using a spectrophotometer or by visual examination due to the bleaching of the hydrogen acceptor. The hill reaction is run in the light and in the dark to demonstrate the light dependence of the reaction.
Photosynthesis II examines the phosphorylation of glucose to produce starch in plant cells. Students collect phosphorylase in potato extract and use it to catalyze the synthesis of starch. They learn that starch synthesis requires primer starch to which glucose residues can be added and that inorganic phosphate inhibits starch synthesis.
Ordering information: This item contains a coupon for perishable materials that must be redeemed from the manufacturer before use. Please allow sufficient time to account for redemption. These materials are shipped from the supplier via second day delivery on Mondays, Tuesdays, or Wednesdays only and must be received promptly.
Delivery information: See datasheet for contents information.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty