Would you like to log in again?
Dental Cavity Potential Lab Activity
Supplier: AQUAPHOENIX SCIENTIFIC INC. SE
8-300
470006-270EA 231.05 USD InStock
470006-270
Dental Cavity Potential Lab Activity
Educational Materials Biology Educational Materials Physiology and Health Learning Activities
Excellent Example of How Microbiology Relates to Physiology
Students will perform a bacterial flora count on their own saliva in this activity designed by faculty from Temple University’s School of Dentistry. They will then perform a test that gives the oral glucose clearance time. After they have completed the activities, students will be able to see what factors play a major role in the formation of cavities. The kit includes enough materials for 24 students, an activity booklet, and student data sheet.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty