Would you like to log in again?
Young's Double Slit Apparatus
Supplier: THE SCIENCE SOURCE SE
22300
470006-476EA 262.15 USD InStock
470006-476
Young's Double Slit Apparatus
Educational Materials Physics Educational Materials Optics Learning Activities
Large scale model of Young's double slit experiment.
  • Adjust the slit spacing and positioning with a simple thumb screw
  • Identify and measure 0th, 1st, and 2nd order bright fringes
  • Teacher's guide included
Demonstrate the wave patterns that create constructive and destructive interference using this large, physical model of Young's Double Slit experiment. Built to scale 20,000 times larger than the actual experimental conditions of a 600 nm HeNe laser illuminating a slit of either 4.5 or 2.25 microns, this apparatus allows calculations to be made for this classical investigation. No lasers or ripple tanks needed for this model - a perfect lab station or lecture aid.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty