Would you like to log in again?
Somso® Animal Cell Model
Supplier: MARCUS SOMMER
ZOS 110/1
470006-482EA 555.3 USD InStock
470006-482
Somso® Animal Cell Model
Models Molecular Biology Models
High-Quality Model Shows Cell Structures In Exquisite Detail
  • Highly Detailed In Bright Colors For Easy Identification
  • Smaller Size Is Ideal For Individual Lab groups Or Stations
  • 5 Year Manufacturers Warranty
View all the major organelles, including the nucleus, endoplasmic reticulum, mitochondria, ribosomes, Golgi apparatus, and centrioles, enlarged 10,000X, on one three-dimensional, colorful model. The one-piece model is mounted on a height-adjustable stand.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty