Would you like to log in again?
Somso® Molar With Cavities Model
Supplier: MARCUS SOMMER
ES 8
470006-486EA 345.95 USD InStock
470006-486
Somso® Molar With Cavities Model
Models Human Biology Models
Recognize The Importance Of Good Dental Hygiene
  • 5 Year Manufacturers Warranty
Perfect for demonstrating the effects of poor oral hygiene, this model comes apart to show a molar, enlarged eight times, with dental cavities detailed from the initial to advanced stages. There are eight different labeled structures, including the tubercles, collum, root and pulp with arteries, veins, and nerves.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty