Would you like to log in again?
Somso® Comparative Germination Model
Supplier: MARCUS SOMMER
BOS 15/7
470006-494EA 1583.45 USD InStock
470006-494
Somso® Comparative Germination Model
Models Botany Models
Three Different Types Of Plants On One Model
  • Angiosperm And Gymnosperm Germination In One Model
  • 5 Year Manufacturers Warranty
Compare monocot, dicot, and gymnosperm germination with one high-quality, exceptionally detailed model. The monocot type is represented by a rye shoot enlarged 10X; the dicot by a two-leaf bean shoot enlarged 5X, and the gymnosperm by a star-shaped fir leaflet enlarged 20X. Students can study root development, the methods by which the different plants sprout, and the structure of each type of seedling because each component of the model is removable for closer examination.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty