Would you like to log in again?
Wildco® Mighty Grab Sampler
Supplier: SCIENCE INTERACTIVE GROUP
78-080
470006-660EA 271 USD InStock
470006-660
Wildco® Mighty Grab Sampler
Samplers Water Samplers
Compact Benthic Dredge with Large Sample Grab
A nearly indispensable tool for sampling hard and soft bottoms, this medium-weight benthic dredge is as easy to use as it is effective. Simply lower the sampler into the body of water and pull up on the included line when it strikes the bottom. You can also use the included sampling pole, which attaches to the trip mechanism, so you avoid getting wet. The durably constructed instrument features stainless steel scoops and a large, easy to handle hinge pin. Advanced features ensure a complete sample — a center pivot closure avoids distortion and the side plates prevent the loss of pebbles. The sampling pole attaches to the trip mechanism for shallow water sampling. Capacity: 1.2 liters. Size: 43 1/2"L.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty