Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ward's® Demonstration Spherical Mirrors

Supplier: G.S.C. INTERNATIONAL, INC.

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Ward's®
4-1301-4 4-1301-3
470006-848EA 88.2 USD InStock
470006-848 470006-846
Ward's® Demonstration Spherical Mirrors
Mirrors
These large silver-backed mirror for demonstrations that can be seen throughout the entire classroom.

  • Large scale mirrors for classroom demonstrations
  • Glass material

The convex mirror weighs 2 lbs., 3.2 oz. The concave mirror weighs 2 lbs., 1 oz. The mirrors come unmounted.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty