Would you like to log in again?
Blood Smearing Kit
Supplier: LAB-AIDS INC (KITS) SE
1B
470007-012KT 74 USD InStock
470007-012
Blood Smearing Kit
Educational Materials Biology Educational Materials Physiology Learning Activities
Students Examine Their Own Blood To Identify Components
  • Learn A Basic Lab Protocol
  • Observe And Identify Different Cell Types
  • Time Required: One 50 Minute Class Period
This three-step experiment provides students with a clear understanding of lab techniques and an opportunity for self discovery. Students draw samples of their own blood with individualized sterile lancets and prepare smears by applying Wright's stain. Using a microscope, they examine the smears to identify different cell types (erythrocytes, polymorphonuclears, eosinophiles, basophiles, lymphocytes and platelets). This experiment can be safely conducted in a single class period.
Included:
Teacher’s Guide with SDS
Wright’s stain, drop controlled bottle
Alcohol pads, pre-saturated
Lancets, sterile, disposable
Materials Needed But Not Supplied: Microscopes, Microscope Slides, Coverslips
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty