Would you like to log in again?
Assorted Plastic Pulleys
Supplier: SI Manufacturing
50030 50030
470007-058EA 30.95 USD InStock
470007-058 470224-022
Assorted Plastic Pulleys
Educational Materials Physics Educational Materials Mechanics Learning Activities
This package of plastic pulleys contains 25 each of 20, 30 and 40 mm diameter pulleys with 3.2 mm bores and one 10 mm pulley with a push fit to a 2 mm motor shaft.
Packaging: Pack of 100.
Caution: CHOKING HAZARD- small parts not for children under 8 years.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty