Would you like to log in again?
Light Crystal Prism
Supplier: TEDCO INC CP
00010
470007-090EA 20.2 USD InStock
470007-090
Light Crystal Prism
Educational Materials Physics Educational Materials Optics Learning Activities
A unique light prism with a flat and slanted edge.
  • Uniquely shaped prism for optics experiments
  • Glass material
Split a beam of light into a multi-colored rainbow with the Light Crystal Prism!
Demonstrate a variety of optic experiments with the Light Crystal Prism - an equilateral prism with a special angled top. Use the enclosed instruction sheet for experiment ideas. The Light Crystal Prism measures approximately 60mm in height, with a 32mm face.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty