Would you like to log in again?
Ward's® Cricket Culture Kit
Supplier: VWR
470007-190KT 84.99 USD InStock
470007-190
Ward's® Cricket Culture Kit
Specimens Live Specimens Animals - Living Material
Study the lifecycle of the brown house cricket (Acheta domesticus) with this complete culture kit ideal for behavioral studies, food for insectivorous animals, or both.
  • Convenient kit allows you to culture your own supply
Students can observe simple metamorphosis at the same time.
It includes 35 crickets, terrarium with cover, substrate, egg-laying mixture, egg-laying dish, water dish, cricket food, and instructions.
Ordering information: This item contains a coupon for live or perishable materials. You will need to provide a date that the live or perishable components will arrive.
Delivery information: The kit will ship separately from the live component. Kits do not contain perishable components and generally ship via UPS ground.
Perishable components will ship via overnight or second day delivery.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty