Would you like to log in again?
Ward's® School Composting Investigations Lab Activity
Supplier: Avantor
WARD470007-200
470007-200KT 279.99 USD InStock
470007-200
Ward's® School Composting Investigations Lab Activity
Educational Classroom Kits and Activities
Involve Your Whole School in Recycling
  • Compost material in an outdoor container
  • Use various carbon sources
  • Includes instructions
Take your indoor composting activities outside by composting waste materials in a large outdoor bin. Teacher’s and students’ manuals are included to guide your class through a variety of investigative projects designed to create an efficient composting system. Along with the bin, you’ll also receive starting inoculum and our comprehensive instructions for setup. You’ll learn how to keep microbes alive so they can continue to degrade your carbon source (leaves, paper, etc.). Size: 33"L x 26"W x 26"H.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty