Would you like to log in again?
Comprehensive Plant Cell Model
Supplier: THE SCIENCE SOURCE SE
B105
470007-358EA 935 USD InStock
470007-358
Comprehensive Plant Cell Model
Models Molecular Biology Models
This translucent model provides a detailed, three-dimensional view of cell.
  • Large Structure for Easy Visibility
  • Clear, Fiberglass-Reinforced Plastic for Enhanced View of Internal Organelles
  • Reference Key Included
  • One-Piece Model
The realistic representation of a plant cell is enlarged approximately 4000 times and includes the nuclear membrane, cell wall, chromosomes, endoplasmic reticulum, chloroplast, large central vacuole, mitochondria, and ribosomes. The model shows 25 organelles and inclusions, and the detailed cell wall shows the relationship to adjacent cells.
Made of translucent fiberglass-reinforced plastic simulating real cytoplasm, this one-piece model is mounted on a base.
Ordering information: The model includes a key identifying 24 structures.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty