Would you like to log in again?
Somso® Deciduous Leaf Section Model
Supplier: MARCUS SOMMER
BOS 17
470007-364EA 587.2 USD InStock
470007-364
Somso® Deciduous Leaf Section Model
Models Botany Models
Comprehensive Review Of A Deciduous Leaf
  • Simplified Structure Of A Deciduous Leaf
  • Key Printed On Base For Easy Reference
  • 5 Year Manufacturers Warranty
This dicot leaf, enlarged 700X, shows the microstructure in fine detail, including a vein, palisade, and parenchyma cells. The cells are sectioned to show the chloroplasts, and the lower surface of the leaf shows the stomate.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty