Would you like to log in again?
Analysis Of Restriction Enzyme Clevage Patterns Of DNA Lab Activity Kit
Supplier: Edvotek
102
470007-374EA 140.75 USD InStock
470007-374
Analysis Of Restriction Enzyme Clevage Patterns Of DNA Lab Activity Kit
Educational Materials Chemistry Educational Materials Chromatography Learning Activities
Restriction enzymes are endonucleases that recognize and cut double-stranded DNA within or near defined base sequences. They are essential tools for biotechnology.
Experiment contains several DNAs digested with various restriction enzymes to demonstrate patterns and frequency of cleavage. Digests are separated by agarose gel electrophoresis.
Include materials for six student groups to run six gels of six lanes each. Requires, but does not include horizontal gel electrophoresis apparatus, DC power supply, and automatic micropipets with tips.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty