Would you like to log in again?
Chemplates®
Supplier: LAB-AIDS INC (KITS) SE
800
470007-402EA 23.95 USD InStock
470007-402
Chemplates®
Microplates PCR Plates
Eliminate the need for glassware and provide a semi-micro environment for conducting many experiments with this clear tray about the size of an index card. You can use Chemplates instead of test tubes, as a demonstration vessel for your overhead projector, for individual student experiments, or even as a gas generator or water reservoir. Twelve large numbered cavities are molded into the high-impact plastic. In addition, each plate has a jumbo-sized cavity and a snap-off measuring spatula. Gas collecting tubes, available separately, fit the Chemplate caps. Package of ten.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty