Would you like to log in again?
Broad Tip Dissecting Forceps
Supplier: DUNRITE INSTRUMENTS SE
140999 141000 140926 141005 141007
470018-958EA 16.75 USD InStock
470018-958 470007-554 470007-552 470018-950 470018-956
Broad Tip Dissecting Forceps
Forceps
Easily hold large sections of tissue.
  • Textured tip for a no slip grip
  • Available in multiple sizes
  • Stainless construction resists rust
Broad-pointed, stainless steel forceps with serrated tips are ideal for coarser dissection work such as handling organs and muscle groups. Large sizes are useful for removing specimens from deep jars, etc. They feature a non-slip grip and come in four sizes.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty