Would you like to log in again?
Helios Planetarium with Northern Hemisphere Stardome
Supplier: COCHRANES OF OXFORD LTD SE
150
470007-594EA 1037.05 USD InStock
470007-594
Helios Planetarium with Northern Hemisphere Stardome
Models Astronomy Models
Helios Planetarium with Northern Hemisphere Stardome.
  • Planets orbit at the correct relative speeds
  • A stardome shows stars for the northern hemisphere
  • Includes a teacher's manual
We consider this among the best instruments you can buy to demonstrate the relative motion of planets around the Sun and the movement of the Moon around Earth.
On the circular base, model planets revolve at their correct relative speeds around a central sun, and the Moon revolves around the Earth. Planets can be positioned for any year and month by means of the degree scale on the rim of the base.
A stardome illustrates the position of the planets among the stars and constellations. The base can also be used to mount the Earth globe and Moon on an inclined axis. When driven, the Earth rotates on its axis, and the Moon revolves around Earth.
The Helios can be used to demonstrate such topics as day and night, seasons, and with the stardome in position, the stars visible at various latitudes.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty