Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

LaMotte® Soil Macronutrients Outfit

Supplier: LA MOTTE CHEMICAL PROD CO SE
LaMotte®
R-5928 R-5928 5928-01
470007-744KT 628.35 USD InStock
470007-744 470199-988 470002-178
LaMotte® Soil Macronutrients Outfit
Standards Environmental Analysis Standards
The individually packaged test factors in this outfit allow several groups to work simultaneously.

  • Spill-Proof Reagent Containers
  • Material for 50 Tests
  • Case Length: 15"
  • Width: 13"
  • Height: 7"

Spill-proof reagent containers hold enough material for 50 tests each of nitrogen, phosphorus, and potassium as well as pH and soil texture.

The 17-pound kit includes instructions and a soil handbook.

Ordering information: It also includes all labware, a soil sampler, instructions, and a soil handbook packed in a lightweight carrying case.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty