Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Metal Storage Cabinets
Supplier: Avantor
1480-MGY 7824-BLK DWR-4218 1480BLACK 7818-MGY DWR-6618 1470-MGY 1470-BLK 7824 7818 J2478SU-MGY J2478SU-BLK
470008-942EA 1042 USD InStock
470008-942 470008-944 470115-678 470115-668 470115-676 470200-582 470115-666 470200-584 470008-938 470200-586 470115-772 470115-770
Metal Storage Cabinets
Furniture Storage Cabinets
Durable metal cabinets provide safe storage for labs and classrooms.
  • Three-point locking mechanism
  • Standard cabinet shelves hold up to 200 lbs.
  • Reinforced doors for strength and quiet operation
  • Adjustable shelving in 2" increments
These sturdy metal storage cabinets are available in five sizes to meet your lab and classroom needs. The cabinets feature two swing-out doors with 3-point locking mechanisms. Interior shelves can be adjusted in 2" increments to provide flexible storage. Storage capacity for Standard and Deluxe cabinets is 200 lbs. and capacity for Jumbo cabinets is 400 lbs.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty