Would you like to log in again?
Kingdom Plantae Microslides
Supplier: SUPERTEK SCIENTIFIC LLC
T-239 T-239-15B
470022-160EA 191.65 USD InStock
470022-160 470009-010
Kingdom Plantae Microslides
Slides Prepared Slides
Investigate plant types and their form and function
  • Arrows help students locate important features
  • Mounted in a clear plastic holder that protects it on both sides
  • Set of 15 includes detailed lesson plan on the topic under study
Introduce junior and senior high school students to the world of microscopy with inexpensive, unbreakable microslides. Microslides consist of a strip of film with eight photomicrographs, each reproduced exactly as you would see them through a professional microscope under ideal conditions. You can supply all your students with their own microslides. A detailed text folder is included. Viewers are available separately. This lesson introduces students to different types of plants.
Contents:
Onion skin (110X), Elodea (150X)
Bean seedling (1/4X)
Moss plant (11/2X); Antheridium (12X); Archegonium (12X)
Fern (4X and 20X)
Female pine cone (21/4X)
Tulip flower (1/5X)
Pistil (5X); Double fertilization (420X)
Monocot seed (20X), Dicot seed (40X)
Box of 15 Sets also Includes:
Background subject information
Reproducible student worksheets with answer keys
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty