Would you like to log in again?
The Digestive System Microslide Set
Supplier: SUPERTEK SCIENTIFIC LLC
T-67-15B T-67
470022-118BX 191.25 USD InStock
470022-118 470009-038
The Digestive System Microslide Set
Slides Prepared Slides
Explore the digestive system up close
  • Arrows help students locate important features
  • Mounted in a clear plastic holder that protects it on both sides
  • Set of 15 includes detailed lesson plan on the topic under study
Introduce junior and senior high school students to the world of microscopy with inexpensive, unbreakable microslides. Microslides consist of a strip of film with eight photomicrographs, each reproduced exactly as you would see them through a professional microscope under ideal conditions. You can supply all your students with their own microslides. A detailed text folder is included. Viewers are available separately.
Contents:
Salivary gland (cs, st, 170X)
Taste buds (ls, st, 350X)
Gullet/esophagus (cs, st, 4X)
Smooth muscle tissue, teased whole cells (st, 700X)
Junction of gullet and stomach (ls, st, 18X)
Stomach lining and gastric glands (cs, st, 85X)
Small intestine (cs, st, 50X)
Intestinal glands and villi (cs, st, 125X)
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty