Would you like to log in again?
Green Plants Microslide
Supplier: SUPERTEK SCIENTIFIC LLC
T-119 T-119-15-B
470022-068BX 191.25 USD InStock
470022-068 470009-098
Green Plants Microslide
Slides Prepared Slides
Examine Different Plants and their Characteristics
  • Arrows help students locate important features
  • Mounted in a clear plastic holder that protects it on both sides
Microslides consist of a strip of film with photomicrographs, each reproduced exactly as you would see them through a professional microscope under ideal conditions. You can supply all your students with their own Microslides. A detailed text folder is included. Viewers are available separately.
Ordering information: Set includes Green leaf (cs, 110X), Green leaf (cs, 350X), Stomate (cs, 480X), Root hairs (wm, 35X), Root (cs, 20X), Celery (Is, 40X), Corn (cs, 145X), Veins in a leaf (wm, 40X), Chloroplasts; The food factory (30,000X), Leaf with starch (wm, 35X), and Bean seed (wm, 2X).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty