Would you like to log in again?
Symbiosis Microslide
Supplier: SUPERTEK SCIENTIFIC LLC
T-212-15B T-212
470009-102PK 12.7 USD InStock
470009-102 470022-078
Symbiosis Microslide
Slides Prepared Slides
Illustrates the Many Interrelationships Between Organisms
  • Arrows help students locate important features
  • Mounted in a clear plastic holder that protects it on both sides
Microslides consist of a strip of film with photomicrographs, each reproduced exactly as you would see them through a professional microscope under ideal conditions. You can supply all your students with their own Microslides. A detailed text folder is included. Viewers are available separately.
Ordering information: Set includes Trypanosomes in rat blood (st, sm, 1,500X), Soybean root nodules (st, cs, 100X), Nematode on tomato root (wm, 200X), Green paramecium (wm, 200X), Rust on willow leaf (st, cs, 200X), Lichen (st, cs, 200X), Fungus on fish (wm, 5X), and Virus on bacteria in man (em, 45,000X).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty